Learn More
Invitrogen™ UQCRC2 Recombinant Rabbit Monoclonal Antibody (24GB2220)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557829
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
UQCRC2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. The core protein 2 is required for the assembly of the complex.
Spezifikation
| UQCRC2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| UQCRC2 | |
| 1500004O06Rik; complex III subunit 2; Core protein II; cytochrome bc-1 complex core protein II; cytochrome b-c1 complex subunit 2, mitochondrial; MC3DN5; QCR2; U MC3DN5; ubiquinol cytochrome c reductase core protein 2; ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II); ubiquinol-cytochrome c reductase core protein II; ubiquinol-cytochrome-c reductase complex core protein 2; UQCR2; Uqcrc2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 293448, 7385 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2220 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22695, P32551 | |
| UQCRC2 | |
| A synthesized peptide derived from human UQCRC2 (aa 300-375). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |