missing translation for 'onlineSavingsMsg'
Learn More

Musashi-2 Antibody (960121) [FITC], Novus Biologicals™

Artikelnummer. 18607653 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18607653 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18607653 Lieferant Novus Biologicals Lieferanten-Nr. FAB3255F

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

Musashi-2 Monoclonal antibody specifically detects Musashi-2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Musashi-2
Anwendungen Flow Cytometry, Immunocytochemistry, CyTOF
Klassifikation Monoclonal
Klon 960121
Konjugat FITC
Verdünnung Flow Cytometry, Immunocytochemistry, CyTOF-ready
Zusammensetzung PBS
Gen-Alias FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2
Wirtsspezies Mouse
Immunogen E. coli-derived recombinant human Musashi-2, Met1-His328, Accession # Q96DH6
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Neuronal Cell Markers
Primär oder sekundär Primary
Gen-ID (Entrez) 124540
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Mehr anzeigen Weniger anzeigen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.