Learn More
Invitrogen™ Ureaplasma parvum Monoclonal Antibody (H71G)
GREENER_CHOICE
Mouse Monoclonal Antibody
Marke:  Invitrogen™ MA517012
Artikelnummer. 15347830
  • 475.00 € / 1 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
MA5-17012 has been used in ELISA and IF applications and shows reactivity with the multi-banded antigen (MB) of several Ureaplasma serotypes. MA5-07012 does not cross react with Mycoplasma hominis and Mycoplasma genitallium. MA5-17012 reacts with serovars: 3 & 14.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Ureaplasma parvum
Monoclonal
0.1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Bacteria
Antibody
IgG1
ELISA, Western Blot, Immunocytochemistry
H71G
Unconjugated
U. parvum
Reactive with the multi-banded antigen (MB) of several Ureaplasma serotypes.
1 mL
Primary
4°C