Spezifikation
| Menge | 0,1 mL |
| Inhalt und Lagerung | Store at -80C. Avoid freeze-thaw cycles. |
| Ziel | USP28 |
| Synonym | Deubiquitinating enzyme 28, EC 3.1.2.15, EC 3.4.19.12, KIAA1515ubiquitin carboxyl-terminal hydrolase 28, ubiquitin specific peptidase 28, ubiquitin specific protease 28, ubiquitin thioesterase 28, Ubiquitin thiolesterase 28, Ubiquitin-specific-processing protease 28 |
| Gensymbol | USP28 |