missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP46 Antibody (CL0363) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-52923PE
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
USP46 Monoclonal antibody specifically detects USP46 in Human samples. It is validated for Western Blot,KnockDown,Immunocytochemistry/ Immunofluorescence
Spezifikation
| USP46 | |
| Monoclonal | |
| PE | |
| Deubiquitinating enzyme 46, EC 3.1.2.15, EC 3.4.19.12, FLJ11850, FLJ12552, FLJ14283, FLJ39393, ubiquitin carboxyl-terminal hydrolase 46, ubiquitin specific peptidase 46, ubiquitin specific protease 46, ubiquitin thioesterase 46, Ubiquitin thiolesterase 46, Ubiquitin-specific-processing protease 46 | |
| This antibody was developed using a recombinant protein derived from P62068, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 64854 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Western Blot, KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0363 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido