missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vaccinia Virus B5 Antibody [Alexa Fluor™ 488] - (Copenhagen), Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Vaccinia Virus B5 Polyclonal antibody specifically detects Vaccinia Virus B5 in Virus samples. It is validated for Western Blot
Spezifikation
Spezifikation
| Antigen | Vaccinia Virus B5 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Alexa Fluor 488 |
| Verdünnung | Western Blot |
| Zusammensetzung | 50 mM Sodium Borate |
| Gen-Alias | OPG190, PS/HR |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Vaccinia virus B5 (Copenhagen) (Uniprot#: P21115; Tyr18-His279) |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?