Learn More
Vaccinia Virus B5 Antibody [FITC] - (Copenhagen), Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Vaccinia Virus B5 Polyclonal antibody specifically detects Vaccinia Virus B5 in Virus samples. It is validated for Western Blot
Spezifikation
Spezifikation
| Antigen | Vaccinia Virus B5 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | FITC |
| Verdünnung | Western Blot |
| Zusammensetzung | PBS |
| Gen-Alias | OPG190, PS/HR |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Vaccinia virus B5 (Copenhagen) (Uniprot#: P21115; Tyr18-His279) |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Mehr anzeigen |