missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAMP1 Polyclonal Antibody
Goat Polyclonal Antibody
Marke: Invitrogen™ PA5148913
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
Synapotobrevins, syntaxins, and the synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Mutations in this gene are associated with autosomal dominant spastic ataxia 1. Multiple alternative splice variants have been described, but the full-length nature of some variants has not been defined.
Spezifikation
| VAMP1 | |
| Polyclonal | |
| Unconjugated | |
| Vamp1 | |
| DKFZp686H12131; lew; MRXSYP; SPAX1; SYB1; Syb-1; synaptobrevin 1; synaptobrevin-1; vamp; Vamp1; VAMP-1; vesicle associated membrane protein 1; vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1) | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 22317, 6843 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P23763, Q62442 | |
| Vamp1 | |
| E. coli-derived recombinant human VAMP-1, Met1-Lys96. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur