Learn More
VAMP4 Mouse anti-Human, CoraLite 488, Clone: 3A6F2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-67219-100UL
Beschreibung
VAMP4 is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family and the SNARE superfamily. Characterized by a common sequence called the SNARE motif, SNARE proteins are involved in membrane fusion and vesicular transport (PMID: 11252968). VAMP4 may play a role in trans-Golgi network to endosome transport.
Spezifikation
| VAMP4 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| IgG2a |
| Immunofluorescence | |
| 3A6F2 | |
| CoraLite 488 | |
| 100 μL | |
| 8674 | |
| Liquid |