Beschreibung
VAPA (Vesicle-associated membrane protein-associated protein A), also known as VAP33, is a 33-kDa, ubiquitously expressed integral membrane protein of the VAMP-associated protein (VAP) family. It is present in the plasma membrane and in intracellular vesicles, playing a role in vesicle trafficking.
Spezifikation
Spezifikation
| Antigen | VAPA |
| Anwendungen | Western Blot, Immunohistochemistry, ELISA |
| Klassifikation | Monoclonal |
| Klon | 1F10F9 |
| Konzentration | 1000 μg/mL |
| Konjugat | Unconjugated |
| Wirtsspezies | Mouse |
| Menge | 150 μL |
| Primär oder sekundär | Primary |
| Gen-ID (Entrez) | 9218 |
| Mehr anzeigen |