missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Varicella zoster Virus glycoprotein E Recombinant Rabbit Monoclonal Antibody (144)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA560396
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Varicella zoster Virus glycoprotein E Recombinant Monoclonal Antibody for ELISA, Western Blot
Varicella Zoster Virus (VZV), a member of the human herpes virus family, causes two distinct clinical manifestations: childhood chickenpox(Varicella) and shingles (zoster). Varicella is the outcome of the primary infection with VZV, whereas, zoster is the result of VZV reactivation from latently infected sensory ganglia which occurs predominantly in aging and immunosuppressed individuals' virons are spherical and 150-200 nm in diameter. Its lipid envelope encloses the nucleocapsid of 162 capsomeres arranged in a hexagonal form. Its DNA is a single linear, double strand molecule, 125,000 nt long.
Spezifikation
| Varicella zoster Virus glycoprotein E | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Recombinant Varicella-zoster virus (VZV)/HHV-3 gE/glycoprotein E protein (Met1-Tyr538). | |
| 20 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 144 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur