Learn More
Invitrogen™ Varicella zoster Virus ORF47 Polyclonal Antibody
Beschreibung
Varicella zoster Virus ORF47 Polyclonal Antibody for Western Blot
The ORF47 gene codes a multifunctional serine/threonine kinase that plays a role in several processes including egress of virus particles from the nucleus, modulation of the actin cytoskeleton and regulation of viral and cellular gene expression. It regulates the nuclear localization of viral envelopment factor proteins 24 and 27, by phosphorylating the protein kinase ORF66, indicating a role in nuclear egress. Disrupts host nuclear lamins, including LMNA and LMNB1. The ORF47 transcript phosphorylates the viral Fc receptor composed of glycoproteins E (gE) and I (gI). Phosphorylation of glycoprotein E (gE) by UL13 alters its subcellular localization, from the host early endosome to the plasma membrane. Participates in the transcriptional regulation of cellular and viral mRNAs mainly by phosphorylating the viral transcriptional regulator IE63.
Spezifikation
Spezifikation
| Antigen | Varicella zoster Virus ORF47 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen-Alias | varicella zoster virus ge |
| Wirtsspezies | Rabbit |
| Immunogen | Synthesized peptide derived from human KR2_VZVD. |
| Reinigungsverfahren | Antigen affinity chromatography |
| Mehr anzeigen |