Learn More
Vasorin/SLIT-like 2 Antibody (101), DyLight 350, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Vasorin/SLIT-like 2 Monoclonal specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Spezifikation
Spezifikation
| Antigen | Vasorin/SLIT-like 2 |
| Anwendungen | ELISA |
| Klassifikation | Monoclonal |
| Klon | 101 |
| Konjugat | DyLight 350 |
| Verdünnung | ELISA, Sandwich ELISA Detection |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin |
| Wirtsspezies | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Vasorin/SLIT-like 2 (Q6EMK4; Met1-Pro575). |
| Mehr anzeigen |