missing translation for 'onlineSavingsMsg'
Learn More

Vat Orange 3 (Technical Grade), TRC

Artikelnummer. 30451785
Change view
Click to view available options
Menge:
5 g
Unit Size:
5 Gramm
Product Code. Menge unitSize
30451785 5 g 5 Gramm
1 options
This item is not returnable. View return policy

Product Code. 30451785

Brand: Toronto Research Chemicals TRCV9950385G

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Vat Orange 3 (Technical Grade)
CAS 4378-61-4
Summenformel C22H8Br2O2
Menge 5 g
InChI-Formel InChI=1S/C22H8Br2O2/c23-15-7-14-19-17-9(15)3-1-5-11(17)21(25)13-8-16(24)10-4-2-6-12(22(14)26)18(10)20(13)19/h1-8H
SMILES Brc1cc2C(=O)c3cccc4c(Br)cc5C(=O)c6cccc1c6c2c5c34
Molekulargewicht (g/mol) 464.11
Formelmasse 461.8891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.