missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human VEGF-165 Recombinant Protein

Artikelnummer. 11858821
Click to view available options
Menge:
20 μg
Packungsgröße:
20 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 11858821

Marke: Invitrogen™ RVEGFI

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Recombinant Protein

Mature human VEGF165 was expressed in Pichia pastoris, KM71. GenBank Accession # 003376, amino acids 27 - 191. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: EGGGQNHHEV. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity of recombinant human VEGF165 was determined by measuring the proliferation of human umbilical vein endothelial cells (HUVEC). Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 1 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.

The vascular endothelial growth factor (VEGF) family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P15692
Zur Verwendung mit (Anwendung) Bioactivity
Zusammensetzung PBS with no preservative
Gen-ID (Entrez) 7422
Molekulargewicht 19.2 kDa
Name Human VEGF-165
Reinigungsverfahren Purified
Menge 20 μg
Quelle Yeast
Lagerungsbedingungen 4°C
Kennzeichnung RUO
Gen-Alias H-VEGF-165; MVCD1; M-VEGF-165; vascular endothelial growth factor 165b; vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; vascular permeability factor; VEGF; VEGF164; VEGF165; Vegfa; VEGF-A; VPF
Gebräuchliche Bezeichnung VEGF-165
Gensymbol Vegfa
Biologische Aktivität Determined by the proliferation effect on human umbilical vein endothelial cells (HUVECs).
Produkttyp Protein
Konjugat Unconjugated
Spezies Yeast
Rekombinant Recombinant
Sequenz Human VEGF-165, amino acids 27-191 (Accession # 003376)
Inhalt und Lagerung 4°C
Aktivität Determined by the proliferation effect on human umbilical vein endothelial cells (HUVECs).
Expressionssystem P. pastoris
Form Lyophilized
Protein-Subtyp Recombinant
Reinheits- oder Qualitätsgrad >95% by SDS-PAGE
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt