Beschreibung
VEGF Monoclonal antibody specifically detects VEGF in Human samples. It is validated for Flow Cytometry, CyTOF-reported
Spezifikation
Spezifikation
| Antigen | VEGF |
| Anwendungen | Flow Cytometry, CyTOF |
| Klassifikation | Monoclonal |
| Klon | 23410 |
| Konjugat | Biotin |
| Verdünnung | Intracellular Staining by Flow Cytometry, CyTOF-reported |
| Zusammensetzung | PBS |
| Gen-Alias | MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor |
| Wirtsspezies | Mouse |
| Immunogen | E. coli-derived recombinant human VEGF |
| Mehr anzeigen |