missing translation for 'onlineSavingsMsg'
Learn More

VEGF Antibody (VEGFA/7758R), Novus Biologicals™

Artikelnummer. p-200004420 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
20 μg
Unit Size:
100 Mikrogramm
20 Mikrogramm
Product Code. Menge unitSize
18641205 20 μg 20 Mikrogramm
18670005 100 μg 100 Mikrogramm
2 options
This item is not returnable. View return policy

Product Code. 18641205

Brand: Novus Biologicals NBP32068120ug

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

VEGF Monoclonal antibody specifically detects VEGF in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF
Anwendungen Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon VEGFA/7758R
Konzentration 0.2 mg/ml
Konjugat Unconjugated
Verdünnung Immunohistochemistry-Paraffin 1-2 μg/mL
Zusammensetzung 10mM PBS with 0.05% BSA
Gen-Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Wirtsspezies Rabbit
Immunogen Recombinant fragment (around aa1-200) of human VEGF protein (exact sequence is proprietary)
Reinigungsverfahren Protein A or G purified
Menge 20 μg
Regulatorischer Status RUO
Forschungsgebiet Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Cytokines and Growth Factors, Developmental Biology, Glycobiology, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision
Primär oder sekundär Primary
Gen-ID (Entrez) 7422
Zielspezies Human
Inhalt und Lagerung Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.