Learn More
VEGF Receptor 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Artikelnummer. 16854885 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
150 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
16854885 150 μL 150 Mikroliter
16864885 20 μL 20 Mikroliter
2 options
Artikelnummer. 16854885
Marke: Proteintech 264151AP150UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
Vascular endothelial growth factor (VEGF) is a major growth factor for endothelial cells. This gene encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of this gene are implicated in infantile capillary hemangiomas.
Spezifikation
Antigen VEGF Receptor 2
Anwendungen Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konzentration 0.4 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gen KDR
Gen-Zugriffsnummer P35918, P35968
Gen-Alias CD309, Fetal liver kinase 1, FLK 1, FLK1, KDR, Kinase insert domain receptor, VEGF Receptor 2, VEGFR, VEGFR 2, VEGFR2
Gensymbole Kdr
Wirtsspezies Rabbit
Immunogen VEGF Receptor 2 Fusion Protein Ag24589
Reinigungsverfahren Antigen Affinity Chromatography
Menge 150 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 16542, 3791
Zielspezies Human, Mouse
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG