Learn More
VGLUT1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 55491-1-AP-150UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16806955
  • 410.00 € / 150 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
Spezifikation
Spezifikation
VGLUT1
Polyclonal
Unconjugated
SLC17A7
BNPI, SLC17A7, VGLUT1
Rabbit
Antigen Affinity Chromatography
RUO
57030
-20°C
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin)
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9P2U7
SLC17A7
Peptide
150 μL