Learn More
Vidarabine, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Vidarabine |
| CAS | 5536-17-4 |
| Summenformel | C10 H13 N5 O4 |
| Menge | 250 mg |
| Synonym | 9H-Purin-6-amine','9-β-D-arabinofuranosyl-, Adenine','9-β-D-arabinofuranosyl- (7CI,8CI), 9-β-D-Arabinofuranosyl-9H-purin-6-amine, 6-Amino-9-β-D-arabinofuranosylpurine, 9-Arabinosyladenine, 9-β-D-Arabinoadenosine, 9-β-D-Arabinofuranosyladenine, 9-β-D-Arabinosyladenine, Adenine 9-β-D-arabinofuranoside, Adenine arabinoside, Adenine β-D-arabinofuranoside, Ara-A, Araadenosine, Arabinosyladenine, Arasena-A, CI-673, NSC 247519, NSC 404241, Spongoadenosine, Vidarabin, Vidarabine, Vidarabine anhydrous, Vira-A, β-Ara-A, β-D-Arabinofuranosyladenine, β-D-Arabinosyladenine |
| InChI-Formel | InChI=1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7+,10-/m1/s1 |
| SMILES | Nc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O |
| IUPAC-Name | (2R,3S,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| Molekulargewicht (g/mol) | 267.24 |
| Formelmasse | 267.0968 |