Learn More
Eppendorf™ Research™ plus BASIC - Trade-In Pipette
Alle ansehen Eppendorf Produkte
Click to view available options
Volumen (metrisch):
0.1 to 2.5 μL
0.25 to 2.5 mL
0.5 to 10 μL
0.5 to 5 mL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Farbe:
Blue
Dark Gray
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Packungsgröße:
1 Stück
Beschreibung
- Volume adjustment feature allows desired volume to be reached in few quick turns
- Control button and ejectors low operating force are positioned for ergonomics 4-digit, magnifying-shape volume display
- Calibration display and adjustment opening for specific liquids and volumes
- Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
- Quick lock makes lower part particularly easy to remove and service
- Spring loaded tip cone promotes tight fit to tip
- Has ability to be autoclaved without disassembling
- Applications: Pipetting liquids, Mixing liquids, Filling of plates, gels, and reaction vessels, Phase extraction and removal of supernatants
Spezifikation
Spezifikation
| Zertifizierung/Konformität | Lot Specific Certificate |
| Kompatible Spitzen | epT.I.P.S. |
| Verstellbarer Spitzenabstand | No |
| Autoklavierbar | Autoclavable |
| Farbe | Medium Gray |
| Einwegprodukt | Reusable |
| Zur Verwendung mit | epT.I.P.S. 20 μL |
| Grifftyp | Ergonomic |
| Material | Plastic |
| Anzahl Kanäle | 1 |
| Mehr anzeigen |