Learn More
Acetylcholinesterase/ACHE Antibody (HR2), DyLight 405 , Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NB300-528V
Artikelnummer. 18702882
  • 668.00 € / 0.20 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beschreibung
Beschreibung
Acetylcholinesterase/ACHE Monoclonal antibody specifically detects Acetylcholinesterase/ACHE in Human, Bovine, Feline, Rabbit, Mouse (Negative), Rat (Negative) samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Acetylcholinesterase/ACHE
Monoclonal
DyLight 405
acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Purified human cerebellar acetylcholinesterase.
0.2 mL
Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
43
Store at 4°C in the dark.
IgG2b
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
HR2
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Bovine, Feline, Rabbit, Mouse (Negative)
Purified