Learn More
Human Ret Antibody, R&D Systems™
Beschreibung
Ret Monoclonal specifically detects Ret in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Spezifikation
Spezifikation
| Antigen | Ret |
| Anwendungen | Western Blot, Flow Cytometry, CyTOF |
| Klassifikation | Monoclonal |
| Klon | 132507 |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gen-Zugriffsnummer | P07949 |
| Gen-Alias | CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence |
| Gensymbole | RET |
| Mehr anzeigen |
For Research Use Only