Learn More
Novus Biologicals™ IFN-alpha/beta R1 Antibody (Medarex patent anti-IFNAR-1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Marke:  Novus Biologicals™ NBP3-28805-50ug
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30045485
  • 213.00 € / 50 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beschreibung
Beschreibung
IFN-alpha/beta R1 Monoclonal antibody specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
IFN-alpha/beta R1
Monoclonal
Unconjugated
P17181
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Medarex patent anti-IFNAR-1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1
IFNAR1
50 μg
Neuroscience, Phospho Specific
3454
Human
Purified