Learn More
Invitrogen™ Human 4E-BP2 (aa 9-38) Control Fragment Recombinant Protein
Artikelnummer. 30211743
Click to view available options
Menge:
100 μl
Packungsgröße:
100 Mikroliter
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30211743
Marke: Invitrogen™ RP103417
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63979 (PA5-63979. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
PHAS-II (eIF4E-binding protein II) is involved in translation initiation. PHAS-II is highest in liver and kidney. PHAS proteins bind to eIF-4E, the mRNA cap-binding protein, and inhibit translation of capped mRNA in vitro and in cells. Phosphorylation in the appropriate site(s) promotes dissociation of PHAS/eIF-4E complexes. This allows eIF-4E to bind to eIF-4G (p220), thereby increasing the amount of the eIF-4F complex and the rate of translation initiation. Pharmacological and genetic evidence indicates that the mTOR/p70S6K pathway is involved in the control of PHAS-I and -II. Thus, PHAS proteins may be mediators of the effects of this pathway on protein synthesis and cell proliferation.
TRUSTED_SUSTAINABILITY
Spezifikation
Zugriffsnummer Q13542
Konzentration ≥5.0 mg/mL
Zur Verwendung mit (Anwendung) Blocking Assay, Control
Zusammensetzung 1 M urea, PBS with no preservative; pH 7.4
Gen-ID (Entrez) 1979
Name Human 4 E-BP2 (aa 9-38) Control Fragment
Menge 100 μl
Kennzeichnung RUO
Gen-Alias 2810011I19Rik; 4 EBP2; 4 E-BP2; AA792569; BC010348; eIF4E-binding protein 2; EIF4EBP2; eukaryotic translation initiation factor 4 E binding protein 2; eukaryotic translation initiation factor 4 E-binding protein 2; PHASII; PHAS-II; Phosphorylated heat- and acid-stable protein regulated by insulin 2; phosphorylated, heat and acid stable regulated by insulin protein II
Gebräuchliche Bezeichnung 4 E-BP2
Gensymbol EIF4EBP2
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung His-ABP-tag
Sequenz HQPSQSRAIPTRTVAISDAAQLPHDYCTTP
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Expressionssystem E. coli
Form Liquid
Reinheits- oder Qualitätsgrad >80% by SDS-PAGE and Coomassie blue staining