missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vimentin Monoclonal Antibody (RV202)
GREENER_CHOICE

Mouse Monoclonal Antibody

Marke:  Invitrogen™ OMA106001

Artikelnummer. 11560193

  • 590.00 € / 100 Mikroliter

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

OMA1-06001 detects vimentin from mouse, rat, hamster, chicken, goat, canine, monkey, zebrafish, porcine and human tissues and cells. OMA1-06001 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, flow cytometry, and immunohistochemistry (frozen) procedures. By Western blot, this antibody detects a ~57 kDa protein representing vimentin from human mesenchymal cells. OMA1-06001 antigen is cytoskeletal vimentin extract of calf lens. Store at 4°C or at -20°C if preferred. Avoid freeze-thaw cycles.

Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
TRUSTED_SUSTAINABILITY
Spezifikation

Spezifikation

Vimentin
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P02543, P08670, P09654, P20152, P31000, P48616, P48670
Vim
Cytoskeletal vimentin extract of calf lens.
100 μL
Primary
Bovine, Canine, Chicken, Goat, Hamster, Human, Mouse, Monkey, Pig, Rat, Frog, Frog
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
RV202
Unconjugated
Vim
cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus
Mouse
Protein G
RUO
100522394, 100689099, 102182515, 140599, 22352, 280955, 420519, 477991, 7431, 81818
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Aktionen

Aktionen

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.