Learn More
Invitrogen™ Vimentin Monoclonal Antibody (VI-RE/1), PE
GREENER_CHOICE
Mouse Monoclonal Antibody
Marke:  Invitrogen™ MA119656
Product Code. 11579202
  • 492.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
This item is not returnable. View return policy
Description
Description
This antibody reacts with human vimentin, a 57 kDa intermediate filament intracellular protein expressed on a wide variety of mesenchymal and mesodermal cell types. This antibody will not cross-react with porcine or mouse.
Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vimentin
Monoclonal
0.1 mg/mL
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P08670
Vim
Bacterially expressed full length human vimentin.
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry
VI-RE/1
PE
Vim
cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus
Mouse
Size-exclusion chromatography
RUO
7431
4°C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications