missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral Tick-borne Encephalitis Virus NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
203.00 € - 456.00 €
Spezifikation
| Antigen | Tick-borne Encephalitis Virus Envelope |
|---|---|
| Klon | 1004120 |
| Verdünnung | Western Blot 1 μg/mL, ELISA |
| Anwendungen | Western Blot, ELISA |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18704287
![]() |
R&D Systems
MAB101061-SP |
25 μg |
203.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18724357
![]() |
R&D Systems
MAB101061-100 |
100 μg |
456.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Tick-borne Encephalitis Virus NS1 Monoclonal specifically detects Tick-borne Encephalitis Virus NS1 in Viral samples. It is validated for Western Blot, ELISA.Spezifikation
| Tick-borne Encephalitis Virus Envelope | |
| Western Blot 1 μg/mL, ELISA | |
| Monoclonal | |
| Ascites | |
| Virus | |
| Human embryonic kidney cell line HEK293-derived viral Tick-borne Encephalitis Virus NS1, Amino Acids 777-1128 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Viral Tick-borne Encephalitis Virus NS1 in direct ELISAs. |
| 1004120 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts