Learn More
Invitrogen™ VISTA Monoclonal Antibody (B7H5DS8), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
Marke: Invitrogen™ 17-1088-42
Beschreibung
Description: This B7H5DS8 monoclonal antibody recognizes human VISTA also known as B7-H5 or PD-H1. This clone has been tested and shown not to cross-react with human B7-H4, B7-H5 and B7-H7. This B7H5DS8 antibody will work in flow cytometry only on live and not paraformaldehyde-fixed cells. Applications Reported: This B7H5DS8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B7H5DS8 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-1H, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans, VISTA is expressed predominantly on hematopoietic cells with the highest densities on myeloid cells, and lower levels on T cells. It is usually absent on B cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. The reported ligands of VISTA include CD28H, VSIG-3 and VSIG-8.
Spezifikation
| VISTA | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9H7M9 | |
| VSIR | |
| VISTA-Fc protein. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| B7H5DS8 | |
| APC | |
| VSIR | |
| 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 64115 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.