Learn More
Vitamin D3 Alexa Fluor 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marke: R&D Systems IC6566G-100UG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Vitamin D3 Monoclonal specifically detects Vitamin D3 in Multi-Species samples. It is validated for Intracellular Staining by Flow Cytometry.
Spezifikation
| Vitamin D3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 25-Hydroxyvitamin D3 | |
| 100 μg | |
| Multi-species | |
| Supernatant |
| Flow Cytometry | |
| 685503 | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |