Learn More
Invitrogen™ Human Vitronectin (VTN) ELISA Kit
Beschreibung
The Human Vitronectin (VTN) ELISA quantitates Hu VTN in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu VTN. Principle of the method The Human VTN solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Spezifikation
Spezifikation
| Zugriffsnummer | P04004 |
| Assay-Bereich | 0,819 ng/mL bis 200 ng/mL |
| Assay-Empfindlichkeit: | 0,8 ng/mL |
| Kalibrierbereich | 0,8 ng/mL bis 200 ng/mL |
| Assay | Festphasen-Sandwich-ELISA |
| Konjugat | Biotin |
| Produkttyp | ELISA-Kit |
| Proteinfamilie | Weitere Proteine |
| Probentyp | Plasma, Serum, Überstand |
| Spezifität | Dieses ELISA-Kit weist keine Kreuzreaktivität mit einem der folgenden getesteten Zytokine auf: humanes A2M, ApoA1, ApoC1, ApoC2, ApoC3, ApoE, ApoH, Artemin, BMP-6, Calbindin D, Ck beta 8-1 , CNTF, EMMPRIN, Endostatin, Fibrinogen, FLRG, GITR Ligand, GPBB, Hämoglobin, HTRA2, IL-17 E, IL-1R3, Lactoferrin, Mesothelin, PAPP-A, PDECGF, PDGF Ralpha, Pepsinogen II, Periostin, Persephin, PTH, RAP, SAA, Serpin F1, Tie-1, tPA, TSP-2, Ubiquitin+1, Vitronectin, vWF |
| Mehr anzeigen |