missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VLDL-Receptor Mouse, Clone: VLDLR/1337, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 7436-MSM1-P0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
| VLDL-Receptor | |
| Cytoplasmic | |
| VLDLR/1337 | |
| Unconjugated | |
| NP_001309154 | |
| Mouse | |
| 143-161kDa | |
| Protein A/G Purified | |
| RUO | |
| 7436 | |
| IgG1 |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| VLDLR | |
| Recombinant human VLDLR fragment from c-terminal | |
| HeLa or U-251 cells. Pancreas, Cerebellum, Heart or Skeletal Muscle. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction