missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Willebrand Factor Antibody (INSERM patent anti-vWF) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Marke: Novus Biologicals (Bio-Techne) NBP3-28369JF525
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spezifikation
| Von Willebrand Factor | |
| Monoclonal | |
| Janelia Fluor 525 | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| vWF | |
| 0.1 mL | |
| Cancer | |
| 7450 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| INSERM patent anti-vWF | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction