missing translation for 'onlineSavingsMsg'
Learn More

Von Willebrand Factor Antibody, Novus Biologicals™

Artikelnummer. 18510621 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.025 mL
Packungsgröße:
25 Mikroliter
Artikelnummer. Menge unitSize
18510621 0.025 mL 25 Mikroliter
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 18510621

Marke: Novus Biologicals NB6005860.025ml

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody has been used in 6 publications

Von Willebrand Factor Polyclonal specifically detects Von Willebrand Factor in Human, Mouse, Canine, Feline samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dual RNAscope ISH-IHC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Von Willebrand Factor
Anwendungen Immunohistochemistry
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:1000
Zusammensetzung 10mM PBS (pH 7.4) with 15mM Sodium Azide
Gen-Zugriffsnummer P04275
Gen-Alias coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Gensymbole VWF
Wirtsspezies Rabbit
Immunogen Purified antisera of the Von Willebrand Factor (factor VIII R:ag) related antigen in plasma.
Reinigungsverfahren IgG purified
Menge 0.025 mL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 7450
Testspezifität The antibody specifically reacts with the cytoplasm of human endothelial cells from normal and neoplastic blood and from lymphatic vessels. It also reacts with human endocardium, platelets and megakaryocytes. It may be used for immunohistological studies of the vascular structures in frozen tissue preparations and enzymatically pre-digested, formalin, methacarn, Bouin's or ethanol-fixed and paraffin-embedded tissue sections.
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.