missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Willebrand Factor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
247.00 €
Spezifikation
| Antigen | Von Willebrand Factor |
|---|---|
| Verdünnung | Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:1000 |
| Anwendungen | Immunohistochemistry |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18510621
|
Novus Biologicals
NB600-586-0.025ml |
0.025 mL |
247.00 €
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Von Willebrand Factor Polyclonal specifically detects Von Willebrand Factor in Human, Mouse, Canine, Feline samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dual RNAscope ISH-IHC.Spezifikation
| Von Willebrand Factor | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P04275 | |
| 7450 | |
| Purified antisera of the Von Willebrand Factor (factor VIII R:ag) related antigen in plasma. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| VWF | |
| IgG | |
| IgG purified | |
| The antibody specifically reacts with the cytoplasm of human endothelial cells from normal and neoplastic blood and from lymphatic vessels. It also reacts with human endocardium, platelets and megakaryocytes. It may be used for immunohistological studies of the vascular structures in frozen tissue preparations and enzymatically pre-digested, formalin, methacarn, Bouin's or ethanol-fixed and paraffin-embedded tissue sections. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts