Learn More
Invitrogen™ Vonlerizumab Recombinant Monoclonal Antibody
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Vonlerizumab, also known as an anti-human CD134 (OX40) monoclonal antibody, is extensively studied for its targeted immunomodulatory function. TNFRSF4, also known as OX40 or CD134, is a gene encoding a protein that belongs to the tumor necrosis factor receptor (TNFR) superfamily. This receptor is primarily expressed on activated CD4 and CD8 T cells and serves as a co-stimulatory molecule involved in the regulation of immune responses. Upon binding with its ligand, OX40L (CD252), OX40 transmits signals that promote T cell proliferation, survival, and cytokine production, thereby enhancing the immune response. The expression of the OX40 gene is tightly controlled and can be upregulated by chromatin remodeling in T cells. OX40 signaling plays a crucial role in the accumulation of protective T cells during anti-viral and anti-tumor immune responses and is being explored as a therapeutic target in both autoimmune diseases and cancer immunotherapy due to its ability to modulate T cell-mediated immune functions.
Spezifikation
Spezifikation
| Antigen | Vonlerizumab Humanized |
| Anwendungen | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Klassifikation | Recombinant Monoclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Wirtsspezies | Human |
| Reinigungsverfahren | Protein A |
| Menge | 100 μg |
| Regulatorischer Status | RUO |
| Mehr anzeigen |