Learn More
Vorolanib, TRC
Artikelnummer. 30451355
Change view
Click to view available options
Menge:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30451355 100 mg
30454418 50 mg
30455430 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30451355 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCV766025100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 1013920-15-4
Summenformel C23H26N5O3F
Molekulargewicht (g/mol) 439.48
Synonym N-[(3S)-1-[(Dimethylamino)carbonyl]-3-pyrrolidinyl]-5-[(Z)-(5-fluoro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-2,4-dimethyl-1H-pyrrole-3-carboxamide, X-82 (Vorolanib)
IUPAC-Name N-[(3S)-1-(dimethylcarbamoyl)pyrrolidin-3-yl]-5-[(Z)-(5-fluoro-2-oxo-1H-indol-3-ylidene)methyl]-2,4-dimethyl-1H-pyrrole-3-carboxamide
SMILES CN(C)C(=O)N1CC[C@@H](C1)NC(=O)c2c(C)[nH]c(\C=C\3/C(=O)Nc4ccc(F)cc34)c2C
Spezifikation
Chemischer Name oder Material Vorolanib
Menge 100 mg
InChI-Formel InChI=1S/C23H26FN5O3/c1-12-19(10-17-16-9-14(24)5-6-18(16)27-21(17)30)25-13(2)20(12)22(31)26-15-7-8-29(11-15)23(32)28(3)4/h5-6,9-10,15,25H,7-8,11H2,1-4H3,(H,26,31)(H,27,30)/b17-10-/t15-/m0/s1
Formelmasse 439.202