missing translation for 'onlineSavingsMsg'
Learn More

Vorolanib, TRC

Product Code. 30455430
Change view
Click to view available options
Menge:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30455430 10 mg
30451355 100 mg
30454418 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455430 Supplier Toronto Research Chemicals Supplier No. TRCV76602510MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013920-15-4
Summenformel C23H26N5O3F
Molekulargewicht (g/mol) 439.48
Synonym N-[(3S)-1-[(Dimethylamino)carbonyl]-3-pyrrolidinyl]-5-[(Z)-(5-fluoro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-2,4-dimethyl-1H-pyrrole-3-carboxamide, X-82 (Vorolanib)
IUPAC-Name N-[(3S)-1-(Dimethylcarbamoyl)Pyrrolidin-3-yl]-5-[(Z)-(5-Fluor-2-oxo-1H-Indol-3-Yliden)Methyl]-2,4-Dimethyl-1H-Pyrrol-3-Carboxamid
SMILES CN(C)C(=O)N1CC[C@@H](C1)NC(=O)c2c(C)[nH]c(\C=C\3/C(=O)Nc4ccc(F)cc34)c2C

Specifications

Chemischer Name oder Material Vorolanib
Menge 10 mg
InChI-Formel InChI=1S/C23H26FN5O3/c1-12-19(10-17-16-9-14(24)5-6-18(16)27-21(17)30)25-13(2)20(12)22(31)26-15-7-8-29(11-15)23(32)28(3)4/h5-6,9-10,15,25H,7-8,11H2,1-4H3,(H,26,31)(H,27,30)/b17-10-/t15-/m0/s1
Formelmasse 439.202
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.