Learn More
Invitrogen™ VPS34 Recombinant Rabbit Monoclonal Antibody (24GB1150)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557604
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
VPS34 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PIK3C3 (hVPS34) is the sole class III member of the PI3 Kinase family. PIK3C3 phosphorylates phosphatidylinositol (PI) to produce phosphatidylinositol 3-phosphate [PI(3)P], which recruits downstream effectors containing FYVE and Phox domains to membranes. PIK3C3 is involved in coupling nutrient signaling to mTOR and also plays a role in autophagy and intracellular trafficking. Involved in regulation of degradative endocytic trafficking and required for the abcission step in cytokinesis, probably in the context of PI3KC3-C2. Involved in the transport of lysosomal enzyme precursors to lysosomes. Required for transport from early to late endosomes.
Spezifikation
| VPS34 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Pik3c3 | |
| 5330434F23Rik; catalytic phosphatidylinositol 3-kinase 3; catalytic subunit type 3 Antibody; class 3 phosphoinositide-3-kinase; Class III PI 3-kinase; hVps34; hVps34 Antibody; OTTMUSP00000023341; OTTMUSP00000023350; phosphatidylinositol 3-kinase; Phosphatidylinositol 3-kinase catalytic subunit type 3; phosphatidylinositol 3-kinase p100 subunit; phosphatidylinositol 3-kinase, catalytic subunit type 3; Phosphoinositide-3-kinase class 3; phosphoinositide-3-kinase, class 3; PI3K type 3; PI-3K Vps34p; pi3kc3; PI3-kinase type 3; Pik3c3; PIK3C3 Antibody; PtdIns-3-kinase type 3; VPS34 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 225326, 5289 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1150 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6PF93, Q8NEB9 | |
| Pik3c3 | |
| A synthesized peptide derived from human PI 3 Kinase Class 3 (aa 800-887). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |