missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WASF3 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 30535377
Change view
Click to view available options
Menge:
100 μg
Artikelnummer. Menge
30535377 100 μg
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30535377

Marke: Invitrogen™ PA5149019

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

WASP (for Wiskott-Aldrich syndrome protein) and N-WASP are downstream effectors of Cdc42 that are implicated in actin polymerization and cytoskeletal organization. The WASP family also includes VASP (vasodilator-stimulated phosphoprotein) and Mena (for mammalian enabled protein), which accumulate at focal adhesions and are also involved in the regulation of the actin cytoskeleton. The WAVE proteins are related to the WASP family proteins and are likewise involved in mediating actin reorganization downstream of the Rho family of small GTPases. The two protein homologs WAVE1 and WAVE2 specifically regulate membrane ruffling by inducing the formation of actin filament clusters in response to GTP binding and activating Rac. The WAVE proteins mediate this actin polymerization by cooperating with the Arp2/3 complex, a nucleation core, and thereby promoting the formation of actin filaments. WAVE1, which is also designated SCAR (for suppressor of cAR), is expressed primarily in the brain, while WAVE2 is widely expressed with the expression highest in peripheral blood leukocytes. WAVE3 forms a multiprotein complex that links receptor kinases and actin. Binding to actin occurs through a C-terminal verprolin homology domain in all family members. The multiprotein complex serves to tranduce signals that involve changes in cell shape, motility or function.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen WASF3
Anwendungen Western Blot
Klassifikation Polyclonal
Konzentration 0.2 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with trehalose and no preservative
Gen WASF3
Gen-Zugriffsnummer Q8VHI6, Q9UPY6
Gen-Alias Brush-1; KIAA0900; Protein WAVE-3; SCAR3; Verprolin homology domain-containing protein 3; WAS protein family member 3; WAS protein family, member 3; Wasf3; WASP family protein member 3; WASP family Verprolin-homologous protein 3; WAVE3; Wiskott-Aldrich syndrome protein family member 3
Gensymbole WASF3
Wirtsspezies Goat
Immunogen E. coli-derived recombinant human WASF3/WAVE3, Met1-Lys180.
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 10810, 245880
Zielspezies Human, Mouse
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Lyophilized
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.