Learn More
Justrite™ Wash Tank with Basket for small parts cleaning,
Product Code. 15300385
Change view
Click to view available options
Höhe (metrisch):
286 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Höhe (metrisch)
15300385 286 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15300385 Supplier Justrite™ Supplier No. 27712
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Typ Wash Tank
Material Stahl
Kapazität (metrisch) 8 L
Höhe (metrisch) 286 mm
Zur Verwendung mit (Geräte) Small Parts Cleaning