missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA541984
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Peptide sequence: TPSLTLQAHS CGINSLHTLP TREGHHLVAS GSEDGSLHVF VLAVEMLQLE Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
WDR6 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation.
Spezifikation
| WDR6 | |
| Polyclonal | |
| Unconjugated | |
| WDR6 | |
| mWDR6; WD repeat domain 6; WD repeat domain 6 protein; WD repeat protein 6; WD repeat-containing protein 6; WDR6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11180 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NNW5 | |
| WDR6 | |
| Synthetic peptide directed towards the C-terminal of human WDR6 (aa 971-1020). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur