Learn More
Invitrogen™ West Nile Virus Envelope (lineage 1, strain NY99) Recombinant Rabbit Monoclonal Antibody (087)
Artikelnummer. 30535461
Change view
Click to view available options
Menge:
20 μL
100 μL
Packungsgröße:
100 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
30535461 20 μL 20 Mikroliter
30535136 100 μL 100 Mikroliter
2 options
Artikelnummer. 30535461
Marke: Invitrogen™ MA560368
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Recombinant Monoclonal Antibody
West Nile Virus Envelope (lineage 1, strain NY99) Recombinant Monoclonal Antibody for Western Blot
West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins which play a major role for WNV entry into target cells. The viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway as delivery of core gene delivery into the striatum of mouse brain and skeletal muscle resulted in cell death and inflammation.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen West Nile Virus Envelope (lineage 1, strain NY99)
Anwendungen Western Blot
Klassifikation Recombinant Monoclonal
Klon 087
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative
Gen-Alias West Nile envelope; West Nile Virus; WNV envelope
Wirtsspezies Rabbit
Immunogen Recombinant West Nile Virus (WNV) (lineage 1, strain NY99) E/Envelope Protein (Domain III) protein (Gln586-Ala705).
Reinigungsverfahren Protein A
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Virus
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG