Learn More
GE Healthcare Optitran™ Membranen aus Nitrocellulose
Beschreibung
- 100% nitrocellulose supported by an inert polyester nonwoven membrane
- Support does not affect transfer conditions or results
- High sensitivity with very low nonspecific binding
- Stringent washing and blocking conditions are not necessary
- Binding capacity: 75 to 90μg/cm2
- Autoclavable using liquid cool cycle
- Also available in circles
Spezifikation
Spezifikation
| Länge (metrisch) | 30 cm |
| Breite (metrisch) | 3 m |
| Bindungsvermögen | 75 to 90 μg/cm2 |
| Menge | Each |
| Format | Rolle |
| Material | Cellulosenitrat |
| Porengröße | 0,45 μm |
| Dimensions (LxW) | 30 cm x 3 m |