Learn More
3M™ White SMS Based Fabric Category III Protective Coveralls
Artikelnummer. 11933764 Alle ansehen 3M Produkte
Ansicht ändern
Click to view available options
Größe:
3X-Large
Packungsgröße:
20 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Größe Packungsgröße
11933764 3X-Large 20 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11933764 Supplier 3M™ Supplier No. 7000089616
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Specifications
Produkttyp Schutzoveralls
Größe 3X-Large
Geschlecht Unisex
Einwegprodukt Single-use
Taillenform Elastisch
Knöchelform Elastisch
Stulpenform Elastic
Flaschenhalstyp Hooded
Haubenausführung 2-teilig
Saumtyp Umgestochen
Sterilität Non-sterile
Schutzartklasse Kategorie III
Höhe (metrisch) 194 bis 200 cm