Learn More
WHSC1 Antibody (CL1057) [Janelia Fluor™ 646], Novus Biologicals™
Product Code. 30291625 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
Product Code. Menge unitSize
30291625 0.1 mL 0.10 Milliliter
1 options
Product Code. 30291625
Brand: Novus Biologicals NBP252962JF646
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
WHSC1 Monoclonal antibody specifically detects WHSC1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Antigen WHSC1
Anwendungen Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Klassifikation Monoclonal
Klon CL1057
Konjugat Janelia Fluor 646
Zusammensetzung 50mM Sodium Borate
Gen-Alias EC 2.1.1.43, FLJ23286, IL5 promoter REII region-binding protein, KIAA1090, MGC176638, MMSETNSD2Multiple myeloma SET domain-containing protein, multiple myeloma SET domain containing protein type III, nuclear receptor binding SET domain protein 2, Nuclear SET domain-containing protein 2, probable histone-lysine N-methyltransferase NSD2, Protein trithorax-5, REIIBP, trithorax/ash1-related protein 5, TRX5, WHS, Wolf-Hirschhorn syndrome candidate 1, Wolf-Hirschhorn syndrome candidate 1 protein
Wirtsspezies Mouse
Immunogen This antibody was developed using a recombinant protein derived from O96028, with the exact immunogen sequence remaining proprietary.
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 7468
Zielspezies Human
Inhalt und Lagerung Store at 4⪚C in the dark.
Form Purified
Isotype IgG1