missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WHSC1 Antibody (CL1063) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-52963CL1
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
WHSC1 Monoclonal antibody specifically detects WHSC1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Spezifikation
| WHSC1 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.1.1.43, FLJ23286, IL5 promoter REII region-binding protein, KIAA1090, MGC176638, MMSETNSD2Multiple myeloma SET domain-containing protein, multiple myeloma SET domain containing protein type III, nuclear receptor binding SET domain protein 2, Nuclear SET domain-containing protein 2, probable histone-lysine N-methyltransferase NSD2, Protein trithorax-5, REIIBP, trithorax/ash1-related protein 5, TRX5, WHS, Wolf-Hirschhorn syndrome candidate 1, Wolf-Hirschhorn syndrome candidate 1 protein | |
| This antibody was developed using a recombinant protein derived from O96028, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL1063 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 7468 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur