missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WIF1 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Marke:  Invitrogen™ PA5148487

Product Code. 30534951

  • 499.00 € / 100 Mikrogramm

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene functions to inhibit WNT proteins, which are extracellular signaling molecules that play a role in embryonic development. This protein contains a WNT inhibitory factor (WIF) domain and five epidermal growth factor (EGF)-like domains, and is thought to be involved in mesoderm segmentation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WIF1
Polyclonal
Unconjugated
WIF1
AW107799; UNQ191/PRO217; WIF; WIF1; WIF-1; Wnt inhibitory factor 1
Goat
Antigen affinity chromatography
RUO
11197
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q9Y5W5
WIF1
Mouse myeloma cell line NS0-derived recombinant human WIF-1, Gly29-Trp379.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.