Learn More
Contec Window Cleaning Squeegee and Replacement Parts
Artikelnummer. p-200132441 Alle ansehen Contec Produkte
Click to view available options
Produkttyp:
Single-blade Squeegee
Packungsgröße:
1 Stück
Artikelnummer. 30280812
Marke: Contec WCSQ0018
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Stainless steel squeegee is lightweight, easy to use, and compatible with QuickConnect handles for hard-to-reach areas. Replacement rubber blade sold separately.
Features Benefits:
  • Lightweight, stainless-steel tool and rubber grip
  • Rubber grip prevents tool from slipping when used with disinfectants
  • Lightweight to clean tall windows and hard to reach areas
  • Maneuvers easily to use vertically and horizontally
Compatibility:
  • Compatible with all Contec's QuickConnect handles making windows easier to reach.
  • All components of the squeegee are compatible with common cleanroom cleaners and disinfectants
Press Down Handle Clip
  • Allows for easy installation preventing the rubber blade from moving during use
Screw-free channel
  • Blades to be replaced quickly and easily saving time
Autoclavable
  • Squeegee can be autoclaved at 121°C for 30 minutes
  • Ideal for use in all areas of the cleanroom
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Single-blade Squeegee
Griffmaterial Stainless Steel
Handle Type Ergonomic
Autoklavierbar Autoklavierbar
Material Gummi