missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WIPF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA554948
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: MQRPSLPDLS RPNTTSSTGM KHSSSAPPPP PPGRRANAPP TPLPMHSSKA PAYNREKPLP PTPGQRLHPG REGPPAPP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.
Spezifikation
| WIPF2 | |
| Polyclonal | |
| Unconjugated | |
| Wipf2 | |
| 1110014J05Rik; 5730509C05Rik; AA407487; Gm1176; PP10631; RGD1561080; WAS/WASL interacting protein family member 2; WAS/WASL interacting protein family, member 2; WAS/WASL-interacting protein family member 2; WASP-binding protein; WASP-interacting protein-related protein; WICH; WIP- and CR16-homologous protein; WIPF2; WIP-related protein; Wire | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 147179 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TF74 | |
| Wipf2 | |
| Recombinant protein corresponding to Human WIPF2. Recombinant protein control fragment (Product #RP-93449). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur